Human Active IL35 (prokaryotic)(from E. coli)
Supplier: CLOUD-CLONE CORP MS
APC008HU01
MSPP-APC008HU1EA 500 USD
MSPP-APC008HU1
Human Active IL35 (prokaryotic)(from E. coli)
Proteins and Peptides
This is a IL35 active protein (prokaryotic), Human is sequencing from Arg21~Lys229+GGGGS+Arg23~Ser219 with 97 to 100% purity. LyopH ilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.
  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee
Interleukin 35 (IL-35) is an IL-12 family cytokine that is produced by regulatory, but not effector, T cells and plays a role in immune suppression. It is a dimeric protein composed of IL-12alpha, and IL-27beta, chains, which are encoded by two separate genes called IL12A and EBI3, respectively. Secreted by regulatory T cells (Tregs), IL-35 suppresses inflammatory responses of immune cells. Studies in mice show the absence of either IL-35 chain from regulatory Tregs reduces the cells ability to suppress inflammation, this has been observed during cell culture experiments and using an experimental model for inflammatory bowel disease. To produce its suppressive effects, IL-35 has selective activities on different T cell subsets, it induces proliferation of Treg cell populations but reduces activity of Th17 cell populations.
Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use In clinical diagnostic procedures. Please proper stored each component based on the instruction.