Soft Temp Reusable Veterinary Pads, Braintree Scientific
Supplier: 0000018294
AP-AV-029 AP-AV-PATCH
MSPP-APAV029EA 81.54 USD
MSPP-APAV029 MSPP-APAVPATCH
Soft Temp Reusable Veterinary Pads, Braintree Scientific
Heaters Hot Plates
Soft-Temp reusable pads are the perfect solution for veterinary warming.
  • Heavy-duty
  • Patched if needed
The pads fit table-tops, surgery tables and in kennel cages. Soft-Temp pads work with Adroit’s HTP-1500 pump, but adapters are available should you have a different pump. Pads may be used directly against the skin or covered with pillow cases or light-weight fabric for added protection against damage.