Human GAS6 active (prokaryotic) (from E.coli)
Supplier: CLOUD-CLONE CORP MS
APA204HU01
MSPP-APA204HU1EA 1472 USD
MSPP-APA204HU1
Human GAS6 active (prokaryotic) (from E.coli)
Proteins and Peptides
This is a GAS6 active protein (prokaryotic), Human is sequencing from Leu136~Phe311 with 90 to 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.
  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee
Growth arrest-specific 6 is similar to the Protein S with the same domain organization and 43% amino acid identity. GAS6 is a gamma-carboxyglutamic acid (Gla) domain-containing protein thought to be involved in the stimulation of cell proliferation. The gene is expressed as an abundant 2.6-kb mRNA in starved cells but decreases significantly upon stimulation of cells with serum or fibroblast growth factor. A human homolog was isolated from HeLa cells and was predicted to encode a 678-amino acid protein with 81% identity to the mouse protein and 44% identity to human protein S, a vitamin K-dependent negative coregulator in the blood coagulation pathway. Regions of greatest similarity between Gas6 and protein S occur in a gamma carboxylated glutamic acid-rich domain and in 4 EGF-like domains.
Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.