Plasmocin™ Treatment - Mycoplasma Elimination Reagent, InvivoGen

Supplier: Invivogen

ANT-MPT
MSPP-ANTMPTEA 340 USD
MSPP-ANTMPT
Plasmocin™ Treatment - Mycoplasma Elimination Reagent, InvivoGen
Cell Culture Supplements and Additives

Plasmocin™ treatment used for the elimination of mycoplasma contamination in cell cultures without affecting cell stemness, pluripotency or viability.


  • Endotoxin level: <2 EU/mg
  • Store at –20 °C


Plasmocin™ is active on both extracellular and intracellular mycoplasmas. It is active at low concentrations on a broad range of gram positive and gram negative bacteria that are otherwise resistant to the mixture of streptomycin and penicillin, and exhibits no toxicity towards eukaryotic cells.


One 1 ml vial is sufficient for 660 ml to 2 liters of culture.


Delivery information: Supplied as a cell culture tested, sterile filtered yellow solution at 25 mg/ml.


Caution: For research use only. Not for human or veterinary use.

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR