HemaPrime™ Biopreservation Cell Freezing Media and Antigens, Antigens and Peptides
Supplier: Charles River Laboratories Cell Solutions
ANTIGEN-1109 ANTIGEN-1095
MSPP-ANTIG1109EA 161.54 USD
MSPP-ANTIG1109 MSPP-ANTIG1095 77529-588 77529-590 77529-592 77529-594 77529-596 77529-598 77529-600 77529-602 77529-604 77529-568 77529-570 77529-572 77529-574 77529-576 77529-578 77529-580 77529-582 77529-584 77529-586
HemaPrime™ Biopreservation Cell Freezing Media and Antigens, Antigens and Peptides
Cell Freezing Media
BioLife solutions and Charles River have partnered to provide CGMP-compliant biopreservation media products. This media significantly extends stability and improves post-preservation viability and function of biologic source material, intermediates, and final products.
Antigen products are proteins and viral lysates that stimulate in vitro immune responses. Various antigens are offered that stimulate immune cells, all of which are tested for bioactivity using PBMCs.
Antigens and peptides: Antigen sequences can be used to evaluate a donors immune response. Peptides are also provided for use with antigen specific T cells and antigen-presenting cells. All antigens are tested for bioactivity using Charles River PBMCs. These antigens are available in 100 μg/vial or 1 mg/vial sizes, depending on the specific antigen.
Delivery information: Dry ice or liquid nitrogen dewar.
Packaging: 1 ml cryovial.
Caution: For research use only. Store at –80 °C until ready to use.