G418 disulfate 100 mg/ml ≥90.0% (by HPLC)

Supplier: Invivogen
Danger

ANT-GN-1
MSPP-ANTGN1EA 153.85 USD
MSPP-ANTGN1
G418 disulfate 100 mg/ml ≥90.0% (by HPLC)
G418 disulphate

G418 is an aminoglycoside antibiotic similar in structure to gentamycin B1 isolated from Micromonospora rhodorangea.


Used for the selection and maintenance of eukaryotic cells expressing the neo gene. This antibiotic blocks polypeptide synthesis by inhibiting the elongation step in both prokaryotic and eukaryotic cells.


Delivery information: G418 is supplied as a sterile filtered solution at 100 mg/ml in HEPES buffer.

Formula: C₂₀H₄₄N₄O₁₈S₂
Storage Temperature: Refrigerator
MDL Number: MFCD00058314
CAS Number: 108321-42-2

Order Now

Specification Test Results

Purity (HPLC) ≥90%
Endotoxin level 0.5 EU/mg
Cell-Culture Tested Potency Validated in G418-Sensitive and G418-Resistant Mammalian Cell Lines
Non-Cytotoxicity of Trace Contaminants Absence of Long-Term Effects Confirmed in G418-Resistant Cells
Concentration 100 mg/ml


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR