Blasticidin, Invivogen
Supplier: Invivogen
ANT-BL-05
MSPP-ANT-BL-05EA 301.54 USD
MSPP-ANT-BL-05
Blasticidin, Invivogen
Microbiology Test Systems Antibiotic Tests
This nucleoside antibiotic specifically inhibits protein synthesis by interfering with the peptidebond formation in the ribosomal machinery in both prokaryotes and eukaryotes. Resistance is conferred by the blasticidin resistance gene, bsr, derived from Bacillus cereus.
  • Endotoxin tested and rigorous quality control
Purity: ≥95%