Human Anti-NPHN ELISA Kit
Supplier: CLOUD-CLONE CORP MS
AEA937HU
MSPP-AEA937HUEA 900 USD
MSPP-AEA937HU
Human Anti-NPHN ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human Anti-NPHN (Nephrin). The assay range is from 3.12 to 200 ng/ml (Competitive kit) with a sensitivity of 1.22 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Nephrin is a member of the immunoglobulin family of cell adhesion molecules that functions in the glomerular filtration barrier in the kidney. The gene is primarily expressed in renal tissues, and the protein is a type-1 transmembrane protein found at the slit diaphragm of glomerular podocytes. The slit diaphragm is thought to function as an ultrafilter to exclude albumin and other plasma macromolecules in the formation of urine. Mutations in this gene result in Finnish-type congenital nephrosis 1, characterized by severe proteinuria and loss of the slit diaphragm and foot processes. Sequence Note: This RefSeq record was created from transcript and genomic sequence data to make the sequence consistent with the reference genome assembly. The genomic coordinates used for the transcript record were based on alignments.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.