Human MBP ELISA Kit
Supplier: CLOUD-CLONE CORP MS
AEA539HU
MSPP-AEA539HUEA 800 USD
MSPP-AEA539HU
Human MBP ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human MBP (Myelin Basic Protein). The assay range is from 3.12 to 200 ng/ml (Competitive kit) with a sensitivity of 1.18 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
Myelin basic protein (MBP) is a protein believed to be important in the process of myelination of nerves in the central nervous system (CNS). Myelin Basic Protein (MBP) is the major constituent of the CNS myelin synthesized by oligodendrocytes and Schwann cells. MBP, PLP (proteolipid proteins) and MOG (myelin oligodendrocyte glycoprotein) are potential primary encephalitogenic target antigens in multiple sclerosis. MBP was initially sequenced in 1979 after isolation from myelin membranes. Since that time, knockout mice deficient in MBP have been developed which showed decreased amounts of CNS myelination and a progressive disorder characterized by tremors, seizures, and early death. The gene for MBP is on chromosome 18, the protein localizes to the CNS and to various cells of the hematopoietic system.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.