Human MOG ELISA Kit
Supplier: CLOUD-CLONE CORP MS
AEA421HU
MSPP-AEA421HUEA 840 USD
MSPP-AEA421HU
Human MOG ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human MOG (Myelin Oligodendrocyte Glycoprotein). The assay range is from 3.12 to 200 ng/ml (Competitive kit) with a sensitivity of 1.21 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
MOG is a glycoprotein believed to be important in the process of myelinization of nerves in the central nervous system (CNS). In humans this protein is encoded by the MOG gene. It’s speculated to serve as a necessary 'adhesion molecule' to provide structural integrity to the myelin sheet and is known to develop late on the oligodendrocyte.
While the primary molecular function of MOG is not yet known, its likely role with the myelin sheath is either in sheet 'completion and/or maintenance'. More specifically, MOG is speculated to be 'necessary' as an 'adhesion molecule' on the myelin sheet of the CNS to provide the structural integrity of the myelin sheet.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.