96 NdFeB ring magnets, grade N48 are designed for use with the SPRI magnetic bead chemistry. Used widely in laboratories and in variety of magnetic bead protocols.
- Recommended reaction volume up to 1000 µl
- Greatly increased microplate compatibility
- Edge effects are minimized because each well in the plate has its own magnet
- More magnetic material per well makes for faster separations
It can be used with 96-well microplates (shallow and half-deep), round-bottom and V-bottom and also with 96-well PCR plates and strips. Integrated spring cushion technology for complete liquid removal without tip occlusion and gripper grooves on the long sides for robotic access.