Clidox®-S Disinfectant, Pharmacal Research Laboratories
Supplier: PHARMACAL RESEARCH LABS MS
96118F
MSPP-96118FEA 25.45 USD
MSPP-96118F
Clidox®-S Disinfectant, Pharmacal Research Laboratories
Cleaning Agents Disinfectants
Clidox®-S is a chlorine dioxide based sterilant . This disinfectant is sporicidal, tuberculocidal, bactericidal, virucidal and fungicidal.
It can be mixed at 1:3:1, 1:4:1 or 1:18:1 . Used to kill both enveloped and non-enveloped viruses.
Clidox-S® diluted 1:3:1 kills bacteria including dried spores of Clostridium sporogenes ATCC 3584 and Bacillus subtillis ATCC 19659 on environmental surfaces in five hours at 25 °C.
Clidox-S® diluted 1:18:1 kills vegetative bacteria including Pseudomonas aeruginosa ATCC 15442, Staphylococcus aureus ATCC 6538, and Salmonella enterica ATCC 10708, pathogenic fungi (Trichophyton mentagrophytes ATCC 9533), viruses, and Mycobacterium bovis ATCC 35743 (tuberculosis bacteria) in five minutes at 20 °C when used on precleaned surfaces.
Clidox-S® diluted 1:4:1 completely inactivates Murine norovirus MNV-1.CW1 in five minutes at 20 °C on pre-cleaned surfaces.