Vessel Caps with TFM Septa, CEM
Supplier: CEM CORPORATION MS
909355
MSPP-909355PK 215.38 USD
MSPP-909355
Vessel Caps with TFM Septa, CEM
Closures Safety Caps and Safety Waste Caps
These caps are compatible with Discover SP-D and Discover SP-D 80 for microwave digestion.