Marprene Tubing, Watson-Marlow
Supplier: WATSON-MARLOW MS
902.0064.016
MSPP-902064016BX 438.46 USD
MSPP-902064016
Marprene Tubing, Watson-Marlow
Tubing Peristaltic Pump Tubing
Marprene tubing is ideal for high chemical resistance to acids and alkalis.
  • Wide chemical compatibility
  • Low gas permeability
  • Opaque to UV and visible light
It offers exceptionally long-life and resistance to a wide range of chemicals, especially oxidizing agents such as ozone, peroxides and sodium hypochlorite.
Operating pressures of 0 to 29 psi can be achieved with Marprene continuous tubing and has an operating temperature of +5 to +80 °C.
Certifications: Meets FDA requirement 21 CFR177.2600 for handling aqueous food and NSF/ANSI 61.