Bacterial Test Standard (BTS), Bruker
Supplier: BRUKER SCIENTIFIC, LLC MS
8255343
MSPP-8255343PK 978.88 USD
MSPP-8255343
Bacterial Test Standard (BTS), Bruker
Controls and Calibration Verification Materials
This bacterial test standard (BTS) is used in the MALDI Biotyper system quality control process to ensure reliable and accurate identification of microorganisms.
Consisting of an E. coli extract spiked with two high-molecular–weight proteins, its composition covers the entire mass range of proteins
measured during MALDI Biotyper identifications.
Delivery information: Box containing five tubes, each sufficient for approximately 40 sample positions.