Tube Rotator and Analog Rotisserie, Scilogex
Supplier: SCILOGEX, LLC MS
82422001
MSPP-82422001EA 671.61 USD
MSPP-82422001
Tube Rotator and Analog Rotisserie, Scilogex
Shakers and Mixers Rotating/Rocking Shakers
MX-RL-E provides a gentle and effective mixing of biological samples in 1.5 to 50 ml micro tubes.
  • Compact bench saving design
  • Circular rotating action
  • Adjustable speed range of 10 to 80 rpm
  • Comes with rotisserie and 36 1.5/2.0 ml and 12ea 15 and 50 ml centrifuge tube clamps
  • Cold room and incubator compatible
  • 2 year warranty
Used in a variety of applications including immune precipitations, prevention of blood coagulation, latex diagnostics and more.