Analog Tube Roller, SCI-T6-S, Scilogex
Supplier: SCILOGEX, LLC MS
823220019999
MSPP-823220019EA 635.38 USD
MSPP-823220019
Analog Tube Roller, SCI-T6-S, Scilogex
Shakers and Mixers Rotating/Rocking Shakers
The SCI-T6-S tube roller provides a gentle but highly efficient rocking and rolling motion.
  • Six roller design for gentle rolling and easy to clean
  • Speed range of 5 - 70 rpm
  • Can be used in cold rooms or incubators
  • DC brushless motor is quiet and maintenance free
  • Protection class IP21
The SCI-T6-S can roll 10×50 ml centrifuge tubes or 10×15 ml centrifuge tubes and many more vessel types. Ideal for mixing blood samples, viscous substances and liquid-solid suspensions.