µ-Slide 18-Well Flat Slide, Ibidi
Supplier: 0000027849
81821
MSPP-81821EA 384.62 USD
MSPP-81821
µ-Slide 18-Well Flat Slide, Ibidi
Slides Chamber Slides
A µ-Slide with 18 wells for use in matrix tests and quick immunofluorescence stainings.
  • Low volume screening slide for medium throughput
  • Cost-effective experiments using small numbers of cells and low volumes of reagents
  • Compatible with multi-channel pipettes
  • Coating area: 0.25 cm²
A convenient multi-well plate for quick and easy tests to probe experimental parameters like antibody dilution, seeding density, or most effective drug concentrations.