sticky-Slide I Luer Slide, Ibidi
Supplier: IBIDI USA MS
81128
MSPP-81128PK 607.69 USD
MSPP-81128
sticky-Slide I Luer Slide, Ibidi
Slides Chamber Slides
A bottomless channel slide used for perfusion applications with a self-adhesive underside designed for perfusion applications and applying defined shear stress and shear rates on cells inside the channel.
  • Channel height: 100 µm
  • Channel volume: 25 µl
  • Channel length: 50 mm
The self-adhesive underside of the bottomless blank slide is easily adapted to a specific bottom substrate.