µ-Slide, 8-Well, Ibidi USA MS
Supplier: 0000027849
80827 80826 80824 80821
MSPP-80827PK 384.62 USD
MSPP-80827 MSPP-80826 MSPP-80824 MSPP-80821
µ-Slide, 8-Well, Ibidi USA MS
Slides Chamber Slides
Designed for high-end microscopic analysis of fixed or living cells.
  • Excellent cell growth on tissue culture treated surface (ibiTreat)
  • Proven mechanical and chemical stability
  • Broad range of sizes and coatings
  • High-resolution microscopy through a coverslip-like bottom
  • 1.0 cm² growth area per well
Ideal for live cell imaging applications like cell cultivation microscopy, transfection, protein localization and high-content microscopy.