Zebra 5095 Thermal Transfer Ribbon, Barcodes Inc.
Supplier: BARCODES INC MS
800132-201
MSPP-800132201CS 59.78 USD
MSPP-800132201
Zebra 5095 Thermal Transfer Ribbon, Barcodes Inc.
Printer Ribbons
The Zebra 5095 performance resin ribbon for printing high-density barcodes and high-resolution graphics on a wide range of materials from matte to gloss synthetics.
This offers exceptional print quality, durability and chemical resistance.
Certifications: UL recognized and CSA component acceptable.
Packaging: 12/carton.