Total RNA Purification Kit
Supplier: STEMCELL Technologies
79040
MSPP-79040EA 644.54 USD
MSPP-79040
Total RNA Purification Kit
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
For purification of RNA from cells.
The Total RNA Purification Kit uses a membrane-based system to quickly purify intact total RNA from 1×10² to 5×10⁶ cells without an organic extraction step, eliminating the need for additional purification steps. The Total RNA Purification Kit includes an on-column DNase I treatment to significantly reduce genomic DNA contamination and ensure the purified RNA is suitable for sensitive downstream applications. Intact total RNA can be isolated in as little as 30 minutes. The Total RNA Purification Kit is not recommended for the purification of small RNAs (<200 nt), such as miRNA.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').