Human/Mouse Recombinant NT-3
Supplier: STEMCELL Technologies
78074.1 78074 78074.2
MSPP-78074-1EA 1476.15 USD
MSPP-78074-1 MSPP-78074 MSPP-780742
Human/Mouse Recombinant NT-3
Proteins and Peptides
Neurotrophin-3 (NT-3) is a neurotrophic factor and a member of the nerve growth factor (NGF) family of proteins that includes neuron growth factor (NGF), brain-derived neurotrophic factor (BDNF), and neurotrophin-4/5. NT-3 signals a number of trophic effects through its transducing receptor tyrosine kinase TrkC. NT-3 is known to promote survival, development, and differentiation of neurons, and modulates transmitter release at several types of synapses in the peripheral and central nervous systems (Chalazonitis 1996). NT-3 has been shown to have an important role in the overall development of enteric neurons, which are crucial for gut peristalsis (Chalazonitis 2004). Studies in rats have shown the potential of NT-3 in dorsal column axonal regeneration (Bradbury et al.). NT-3 was shown to protect neurons against amyloid-β toxicity (Lesne et al.). NT-3 has applications in neuronal differentiation protocols to generate β-tubulin III+ peripheral neurons from neural crest stem cells (Menendez et al.) and oligodendrocyte precursor cells from human embryonic stem (ES) and induced pluripotent stem (iPS) cells (Douvaras et al.).
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').