Magnetic Bead Separation Blocks, 384 Well Microplates, V&P SCIENTIFIC INC MS
Supplier: V & P SCIENTIFIC INC MS
VP771G-4AAZM-1
MSPP-771G4AAM1EA 2217.83 USD
MSPP-771G4AAM1
Magnetic Bead Separation Blocks, 384 Well Microplates, V&P SCIENTIFIC INC MS
Racks Magnetic Separation Racks
Designed for robotic gripper access for microplate movement.
  • Red anodized aluminum frame
  • Zinc coated steel plate
  • 425 post 52 MGO neodymium magnet
  • SBS footprint
Magnetic bead separation block is used for 384 well microplates with flat bottom wells only.
Bead pellets are located in the 4 corners/sides, bottom.
North poles up, axial magnetic orientation.
Delivery information: Includes anodized aluminum registration base.