Trypan blue 0.4% in PBS

Supplier: STEMCELL Technologies
Danger

Synonyms: Tolidine-disazo-bis-8-amino-1-naphthol-3,6-disulphonic acid tetrasodium salt, CI 23850, Tolidine-disazo-bis-8-amino-1-naphthol-3,6-disulfonic acid tetrasodium salt, 3,3′-Dimethyl-4,4′-bis(5-amino-4-hydroxy-2,7-disulphonaphthyl-3-azo)-[1,1′-biphenyl] tetrasodium salt, Benzamine blue, Trypan blue sodium salt, Direct Blue 14, Diphenyl Blue, Sodium 3,3'-((3,3'-dimethyl-[1,1'-biphenyl]-4,4'-diyl)bis(diazene-2,1-diyl))bis(5-amino-4-hydroxynaphthalene-2,7-disulfonate), AzidinBlue, 3,3′-Dimethyl-4,4′-bis(5-amino-4-hydroxy-2,7-disulfonaphthyl-3-azo)-[1,1′-biphenyl] tetrasodium salt, Sodium 3,3'-((3,3'-dimethyl-[1,1'-biphenyl]-4,4'-diyl)bis(diazene-2,1-diyl))bis(5-amino-4-hydroxynaphthalene-2,7-disulphonate)

07050
MSPP-70-50EA 102.31 USD
MSPP-70-50
Trypan blue 0.4% in PBS
Trypan Blue

Reagent for counting viable mammalian cells.


Trypan Blue is recommended for counting viable mammalian cells. Viable cell counts should be performed where a decrease in cell viability may be expected, for example, when working with cryopreserved cells or cells manipulated ex vivo.


Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Formula: C₃₄H₂₄N₆Na₄O₁₄S₄
MW: 960.82 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003969
CAS Number: 72-57-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR