Reagent Reservoirs, 12 Column, Integra Biosciences
Supplier: INTEGRA MS
6361
MSPP-6361CS 301.54 USD
MSPP-6361
Reagent Reservoirs, 12 Column, Integra Biosciences
Reagent Reservoirs
The polypropylene design improves the reservoirs chemical compatibility and also allows them to be autoclaved.
  • Reagent reservoirs for 96 well plates
  • Pyramid bottom
  • Each column holds 21 ml