Anti-CD3 Mouse Monoclonal Antibody [clone: UCHT1] (PE)

Supplier: STEMCELL Technologies
100-0449 60011PE 60011PE.1
MSPP-100-0449EA 1946.77 USD
MSPP-100-0449 MSPP-60011PE MSPP-60011PE1
Anti-CD3 Mouse Monoclonal Antibody [clone: UCHT1] (PE)
Antibodies
Mouse monoclonal IgG1 antibody against human, chimpanzee CD3, PE-conjugated.

  • T cells

The UCHT1 antibody reacts with the ~20 kDa CD3ε subunit of the human T cell receptor (TCR)/CD3 complex, which is expressed on the surface of ~95% of mature T cells and NKT cells, and variably on thymocytes. A majority of T cell neoplasms also express CD3. The CD3 complex, which is assembled from combinations of CD3γ, δ, ε, η, and ζ subunits, associates non-covalently with the TCR and is involved in transducing antigen recognition signals into the cytoplasm of T cells and in regulating the cell surface expression of the TCR. Activation of T cells by the TCR involves the cytoplasmic tails of the CD3 subunits, which are structurally related type 1 transmembrane proteins and members of the immunoglobulin superfamily. Mutations in the CD3 subunits have been associated with various immunodeficiency disorders including severe combined immunodeficiency (SCID). This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Direct Human T Cell Isolation Kit (Catalog #19661), EasySep™ Human CD3 Positive Selection Kit II (Catalog #17851; partial blocking may be observed), EasySep™ HLA Whole Blood T Cell Enrichment Kit (Catalog #19951HLA) and EasySep™ HLA Whole Blood CD2 Positive Selection Kit (Catalog #18687HLA).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: CD3
Clonality: Monoclonal
Clone: UCHT1
Conjugation: PE
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Chimpanzee;Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR