Hemocytometer, Disposable, Diagnocine
Supplier: DIAGNOCINE LLC MS
FNK-521-10-1
MSPP-52110PK 214.29 USD
MSPP-52110
Hemocytometer, Disposable, Diagnocine
Slides Counting Chambers
Disposable hemocytometer with 4 chambers, made of plastic.
  • Designed for rapid loading
  • No washing, no labor, saves time
  • Same grid as standard hemocytometer
  • High calculation accuracy