BTXpress Cytofusion® Electroporation Medium C, Harvard Apparatus

Supplier: Harvard Apparatus
47-0001
MSPP-47-0001EA 606.92 USD
MSPP-47-0001
BTXpress Cytofusion® Electroporation Medium C, Harvard Apparatus
Electroporation Systems Electroporation Kits
BTXpress Cytofusion® Medium C is an advanced electrofusion buffer designed for use with the BTX Hybrimune System for high performance cell fusion applications.

  • Osmolarity: 270 to 290 mOsm/l
  • Conductivity at 25 °C: 0.080 ±0.005 mS/cm
  • pH: 7.2 ±0.2
  • Endotoxin: <0.25 EU/ml

The low conductivity buffer is specially-formulated to minimize cell turbulence during cell alignment and heating during electrofusion for robust cell fusion efficiency and high cell viability. It is sterile filtered from the highest quality non-animal, medical-grade reagents. It is the buffer of choice for many commercial biotech and pharmaceutical companies in their hybridoma generation process for monoclonal antibody discovery.

It is sterile filtered and optimized for eukaryotic electrofusion applications and also has stable environment for cell alignment.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR