CellTrics® Disposable Cell Strainers, Sysmex Amercica
Supplier: SYSMEX AMERICA, INC MS
04-004-2327
MSPP-40042327PK 168.62 USD
MSPP-40042327
CellTrics® Disposable Cell Strainers, Sysmex Amercica
Cell Strainers
CellTrics® are disposable filters for isolation of cells and nuclei from cell debris and aggregates.
  • Not autoclavable
  • One-time use only
  • No additional devices are required
  • 5 µm mesh diameter
These sterile filters are single-packed and have tilted filters made from monofil nylon material.
Caution: For research use only.