Stablcal® Turbidity Standards
Supplier: HACH COMPANY MS
2896742 2896842 2897042 2896942 2660142 2660149 2660242 2660249 2660442 2660449 2660542 2660549 2659742 2659749 2659753 2659842 2659849 2659853 2659942 2746353 2697942 2697953 2698042 2723342 2723353 2659949 2660153 2660642 2660649
MSPP-2896742EA 105.57 USD
MSPP-2896742 MSPP-2896842 MSPP-2897042 MSPP-2896942 104086-540 104086-542 104086-544 104086-546 104086-548 104086-550 104086-552 104086-554 104086-526 104086-528 104086-530 104086-532 104086-534 104086-536 104086-538 104086-620 104086-582 104086-584 104086-586 104086-600 104086-602 104088-268 104088-270 104088-272 104088-274
Stablcal® Turbidity Standards
Standards Turbidity Standards
Stablcal® turbidity standards for all turbidimeters. USEPA accepted for calibration purposes. Stablcal® stabilized formazine turbidity standards are true formazine dilutions developed for use in any turbidimeter.
  • Precise, pre-mixed formazine solutions
  • Based on formazine primary standards
  • Enhanced stability saves time and labour
  • Confidence in results