Self Piercing Ear Tags, Fine Science Tools
Supplier: FINE SCIENCE TOOLS INC MS
24220-10
MSPP-2422010EA 53.08 USD
MSPP-2422010
Self Piercing Ear Tags, Fine Science Tools
Animal Supplies Animal ID Tags and Punches
Ideal for use in small animals such as mice and rats, these ear tags come numbered from 1 to100.
Silver colored tags with black numbers provide high visibility making them easy to read. Once applied, tags seal and cannot be removed, providing an easy to use, permanent identification method. These ear tags are safe for use in salt water.