Wipe Dispenser for Kimwipes®, Labscientific
Supplier: LAB SCIENTIFIC BIOKEMIX, INC. MS
234522M
MSPP-234522MEA 65.11 USD
MSPP-234522M
Wipe Dispenser for Kimwipes®, Labscientific
Wipe Dispensers
Wipe dispenser holds packages large lab wipes. The smaller version attaches with double-back tape and the larger version with screws.
  • Mount on a shelf or wall
  • Fit Kimwipes® and other brands
Delivery information: Mounting materials are included.