Cell Lifters, Celltreat®
Supplier: CELLTREAT SCIENTIFIC PRODUCTS MS
229306
MSPP-229306CS 162 USD
MSPP-229306
Cell Lifters, Celltreat®
Cell Scrapers
Unique double ended cell lifters features a 19 mm beveled edge flat blade on one end, and J-hook on the other.
  • Flat blade ensures even contact with growth surface of flasks and dishes
J-hook is curved on one end to vertically 4, 6, 12, 24, and 48 well plates.
Packaging: Individually wrapped in bag.