Sulphite Standard Solution
Supplier: Hach
2267410
MSPP-2267410EA 139.85 USD
MSPP-2267410
Sulphite Standard Solution
Standards Environmental Analysis Standards
This equivalent standard is prepared with stabilized sodium thiosulfate, since sulphite is not a stable in solution.