EasySep™ Buffer
Supplier: STEMCELL Technologies
20144
MSPP-20144EA 171 USD
MSPP-20144
EasySep™ Buffer
Cell Separation Media
EasySep™ Buffer is recommended for EasySep™ cell separation protocols.
  • Can be used in chimerism, HLA, immunology
Species: Human, mouse, non human primate, rat and others.
Delivery information: Supplied with Dulbecco's phosphate-buffered saline (PBS), Fetal bovine serum (2%) and EDTA (1 mM) in PBS.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').