Hausser Scientific™ Hemocytometer Cover Glasses
Supplier: STEMCELL Technologies
200-0641
MSPP-200-0641EA 295.23 USD
MSPP-200-0641
Hausser Scientific™ Hemocytometer Cover Glasses
Cover Glasses
Count cells with hemocytometer cover glasses designed specifically for use with the Hausser Scientific™ Bright-Line Hemocytometer.
  • Replacement cover glasses
These cover glasses facilitate manual cell counting at a fraction of the cost and footprint of automated cell counters.
Packaging: They are available in packs of 10.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').