EasySep™ Direct Human B-CLL Cell Isolation Kit
Supplier: STEMCELL Technologies
19664
MSPP-19664EA 1075.38 USD
MSPP-19664
EasySep™ Direct Human B-CLL Cell Isolation Kit
Cell Separation Media
Immunomagnetic isolation of B-CLL cells directly from whole blood
  • >99.9% RBC depletion without the need for density gradient centrifugation, sedimentation, or lysis
  • Fast, easy-to-use and column-free
  • Isolated cells are untouched
This kit isolates untouched and highly purified B-CLL cells directly from human whole blood by fully immunomagnetic negative selection. The depletion cocktail in this kit does not contain antibodies against CD43, which can be expressed on malignant cells in chronic lymphocytic leukemia (CLL) samples.
This cell isolation kit targets non-B cells for removal based on specific surface antigens. Unwanted cells are labeled with antibodies and EasySep™ Direct RapidSpheres™, and separated using an EasySep™ magnet. Desired cells are simply collected into a new tube and can immediately be used in downstream applications such as nucleic acid extraction, flow cytometry or culture.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').