Density Analyzer, Autotap Dual Station
Supplier: ANTON PAAR USA MS
192980
MSPP-192980EA 9815.38 USD
MSPP-192980
Density Analyzer, Autotap Dual Station
Analyzers Elemental Analysers
The Dual Autotap tapped density analyzer for powders is simple-to-use, automated device which comply with a large number of internationally recognized standard methods.
  • Two automatically rotating tapping stations
  • 260 taps per minute
  • 3 mm drop height
  • Ensure the quality of raw materials and predict the flow behavior of powders
  • Accommodates a wide range of graduated cylinder sizes for full flexibility
  • Operates in accordance with a large number of international standard test methods
It provides a high level of test method control with a user-selectable, lockable number of taps. Tapping rate and drop height are fixed to provide consistent results from sample to sample. A large range of sample sizes can be accommodated with the use of a number of different graduated cylinder sizes.
Dual Autotaps has been used to provide customers with consistent, reliable bulk density and flowability values – essential knowledge for measuring pharmaceutical powders, metal powders, food powders, pigments, and more.
Dual Autotaps accommodate any single-piece glass-graduated cylinder with a hexagonal base. The tapping platforms have hole patterns and adjustable hold-down clamps so that any graduated cylinder from 5 to 500 ml may be used. There is also an adapter should you need to measure in a 1 L graduated cylinder.
The design of Dual Autotaps ensures compliance with all internationally recognized standard test methods describing the use of tapped density to determine bulk density involving a device capable of ensuring a drop height of 3 mm, a tapping frequency between 250 and 300 taps per minute, and the use of a specific type of graduated cylinder.