Octet® Protein A (ProA) Biosensor, Sartorius
Supplier: SARTORIUS CORPORATION MS
18-5012
MSPP-185012PK 5046.15 USD
MSPP-185012
Octet® Protein A (ProA) Biosensor, Sartorius
Assays Protein Assays
For determination of antibody concentration.
  • Direct measurement of immunoglobulins (IgG)
  • Assay samples without centrifugation
  • Fast turnaround of results
  • Broad dynamic range
  • Correlates to HPLC
  • Dynamic range 1–500 μg/ml for most proteins
Throughput: 8 samples in about 2 minutes, 96 samples in about 32 minutes.
Ordering information: Additional materials are required to run these assays. Recommended sample diluent sold separately. Standards: A purified standard that is identical to the experimental samples is required. Media for sensor hydration: the biosensor hydration solution must match the standard and experimental sample matrix (e.g., blank media or sample diluent). Contact your Avantor representative for more information.
Caution: For research use only.