EasyEights™ EasySep™ Magnet
Supplier: STEMCELL Technologies
18103
MSPP-18103EA 3585.15 USD
MSPP-18103
EasyEights™ EasySep™ Magnet
Magnets
The EasyEights™ EasySep™ Magnet is designed for cell separation procedures using selected EasySep™ reagents when simultaneously processing multiple samples, up to eight samples on each side, for a total of 16 samples.
  • Magnet for column-free immunomagnetic separation
The EasyEights™ EasySep™ magnet has a large sample range, capable of processing small samples, starting at 0.1×10⁸ cells, to larger samples, up to 8.5×10⁸ cells. The EasyEights™ EasySep™ magnet is designed to hold up to 8 standard 5 ml (12×75 mm) round-bottom tubes on one side, and 8 standard 14 ml (17×95 mm) round-bottom or 15 ml conical tubes on the other side.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').