EasySep™ Magnet
Supplier: STEMCELL Technologies
18000
MSPP-18000EA 1780.15 USD
MSPP-18000
EasySep™ Magnet
Magnets
The EasySep™ magnet is designed for cell separation procedures using EasySep™ reagents.
  • Immunomagnetic column-free magnet
  • Designed to hold a standard 12×75 mm (5 ml) polystyrene tube
The EasySep™ magnet generates a high-gradient magnetic field in the interior cavity that is strong enough to separate cells labeled with EasySep™ magnetic particles, without the use of columns.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').