EasySep™ Human PSC-Derived Cardiomyocyte Enrichment Kit
Supplier: STEMCELL Technologies
17965
MSPP-17965EA 784.62 USD
MSPP-17965
EasySep™ Human PSC-Derived Cardiomyocyte Enrichment Kit
Cell Separation Media
Immunomagnetic negative selection kit.
  • Fast and easy-to-use
  • Up to 99% purity
  • Isolated cells are untouched
The EasySep™ Human PSC-Derived Cardiomyocyte Enrichment Kit is designed to isolate human PSC-derived cardiomyocytes from cell cultures by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-cardiomyocytes and dextran-coated magnetic particles. The labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).