EasySep™ Human Neutrophil Isolation Kits
Supplier: STEMCELL Technologies
17957 17957RF
MSPP-17957EA 1898.46 USD
MSPP-17957 MSPP-17957RF
EasySep™ Human Neutrophil Isolation Kits
Cell Separation Media
Immunomagnetic isolation of untouched human neutrophils.
  • Fast, easy-to-use and column-free
  • Up to 99% purity
  • Untouched, viable cells
The EasySep™ Human Neutrophil Isolation Kit is designed to isolate neutrophils from a polymorphonuclear cell-rich fraction of peripheral blood by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-neutrophils and magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube. The isolation kit is compatible with cells prepared using HetaSep™ sedimentation or red blood cell lysis.
This product replaces the EasySep™ Human Neutrophil Enrichment Kit, for even faster cell isolations.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').