ALDHbr Assay Kit
Supplier: STEMCELL Technologies
01711
MSPP-1711EA 2648.31 USD
MSPP-1711
ALDHbr Assay Kit
Assays Cellular Assays
For the detection of CD34+ and ALDHbr cells in human cord blood –6×10⁷ cells.
  • Quantifies primitive HSCs in <1 day
  • Optimized to detect ALDHbrCD34+ cells in CB
  • Enumerates viable, metabolically active, primitive cells in the CD34+ cell population
The ALDHbr Assay Kit is optimized for the identification and quantitation of CD34+ cells that express high levels of the enzyme aldehyde dehydrogenase (ALDH) in human cord blood samples. This kit contains both the CD34+ Cell Detection Kit and the ALDEFLUOR™ Kit. The CD34+ Cell Detection Kit contains pre-selected fluorochrome-conjugated monoclonal antibodies that have been optimized for use with the ALDEFLUOR™ Kit to detect cells that express CD34 and have high ALDH activity by flow cytometry. The CD34+ Cell Detection Kit contains anti-human antibodies APC CD34, PE CD45, and PE-Cyanine5 Glycophorin A/B, and 7-AAD viability dye.
A flow cytometer equipped with a 488 nm laser and a 635 nm laser for excitation and appropriate filters for detecting APC, PE, PE-Cyanine5 and 7-AAD fluorescence is required for use.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').