RosetteSep™ DM-M Density Medium
Supplier: STEMCELL Technologies
15725
MSPP-15725EA 168.08 USD
MSPP-15725
RosetteSep™ DM-M Density Medium
Cell Separation Media
Density gradient medium for use with RosetteSep™ myeloid cell enrichment cocktail.
  • RosetteSep™ DM-M has a density of 1.085 g/ml
  • For use in cell isolation, In Vitro diagnostic applications
RosetteSep™ DM-M is a density gradient medium designed specifically for use with RosetteSep™ cocktails for the enrichment of specific human myeloid (CD33+) cells from whole blood.
Caution: In light of the new EU Regulation 2017/746 (IVDR) of the European Parliament and of the Council on in vitro diagnostic medical devices, which is expected to become effective in 2022, STEMCELL recently deregistered and removed the CE IVD claims associated with these products, and they are now labeled 'For Research Use Only'. Please note there have been no changes to the form, function and quality assurances associated with these products. For more information please contact the STEMCELL Technologies Quality Assurance and Regulatory Department.
Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet ( collectively, the ‘Product Restrictions’).