Sulfuric acid 1.600 N, cartridge for digital titrator

Supplier: Hach
Danger

1438901
97026-514EA 57.6 USD
97026-514 MSPP-1438901
Sulfuric acid 1.600 N, cartridge for digital titrator
Sulfuric acid

For determination of alkalinity by Digital Titrator titration.


  • Accurate results when and where you need them
  • Fast and Flexible
  • Versatile
  • Economical


Hach method 8203.
1.600 N Sulfuric Acid. Digits = mg/L CaCO3 in 100 mL, Digits x 0.002 = meq.

Formula: H₂SO₄
MW: 98.08
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.84
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
UN: 3264
ADR: 8 II

Order Now

Specification Test Results

EPA compliant Yes
Method Punto final de fenolftaleína
Method Name Phenolphthalein
Method Number 1 Chemistry 8203
Number of tests 40 - 100
Parameter Alcalinidad, base ácida
Platform Digital Titrator
Range 100 - 4000 mg/L CaCO3
Range 2 1 - 10 meq/L Base
Titrant 1.600 N H2SO4


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR