Artery Scissor, Fine Science Tools
Supplier: FINE SCIENCE TOOLS INC MS
14080-11
MSPP-1408011EA 169.23 USD
MSPP-1408011
Artery Scissor, Fine Science Tools
Scissors
Scissor with one ball tip to prevent piercing of underlying tissue.
  • Not serrated
  • With 1.5 mm ball diameter
Cutting edge: 25 mm