Temperature Monitor with Alarm, Single Sensor, Model T-Sentry 140, Hampshire Controls Corp MS
Supplier: HAMPSHIRE CONTROLS CORP MS
F82040-20
MSPP-140-100EA 744.62 USD
MSPP-140-100
Temperature Monitor with Alarm, Single Sensor, Model T-Sentry 140, Hampshire Controls Corp MS
Monitoring Systems CO2 and Temperature Monitoring Systems
The Model T-Sentry 140 is a stand-alone continuous single-probe temperature monitoring system with alarm status notification system.
  • Single temperature sensor
  • Easy to read LCD display
  • Audible and visual alarms
  • Adjustable high/low alarm settings and alarm delay period
  • Selectable alarm mute period
  • Standard operating configuration (-100...+100 °C)
  • Optional Email notification of alarm conditions
  • Optional Ethernet integration into Model AMS monitoring, logging and alarm status notification system
It is widely used within refrigerators, freezers and walk-in coolers - wherever material storage temperatures must be maintained within a continuously tight range. Standard sensors 100 ohm RTD in 4.0" or a 2.0" stainless steel sheath with 10' of lead wire.
By selecting a different probe type/temperature range the system can also be used for high temperature monitoring applications (e.g., within ovens, heating chambers and other manufacturing machinery).
Includes user-settable high/low temperature alarm indication thresholds, audible/visual alarms (via beeper and LEDs), programmable alarm mute and 'chirp' periods and a UL-listed plug-in power supply.
Additional features include audible alarm muting (for user-selectable periods up to 32 minutes), optional relay contacts (for indicating temperature and/or power failure conditions). and an optional alarm triggered auto dialing notification system. The system is intuitive and easily programmed for any application.
Ordering information: For all available options (sold separately), contact your Avantor representative for more information.