CryoSealer, Manual Ultrasonic Sealer for Straws
Supplier: MINITUBE USA, INC. MS
131350000
MSPP-131350000EA 6433.54 USD
MSPP-131350000
CryoSealer, Manual Ultrasonic Sealer for Straws
Heat Sealers Bag Sealers
Ultrasound welding facilitates a perfect and uniform seal of each straw.
  • Focused heat generation instead of undefined temperature radiation
  • Defined straw position (fixed angle and depth of insertion)
  • Sealing unit closes fully automatic
  • Standardized welding pressure due to preset welding gap
  • Precise and reproducible selection of the welding time based on the operating instructions
This cryosealer can be used for 0.25, 0.5 ml straws and 0.3 ml CBS cryobiostraws.